ENA321763069 Enamine

2-[3-(3-phenylpropoxy)phenyl]acetonitrile

  • MDL number MFCD18785502
  • Cas Number : 1311317-75-5
  • Molecular Weight : 251.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74985

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74985
Availability details for
Quantity
  • Availability
  • Add to Cart