ENA321763072 Enamine

tert-butyl 4,4-dimethyl-2,5-dioxoimidazolidine-1-carboxylate

  • MDL number MFCD18785505
  • Cas Number : 1311315-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75011
Availability details for
Quantity
  • Availability
  • Add to Cart