ENA321763073 Enamine

2-chloro-N-{4-[(pyrrolidin-1-yl)methyl]-1,3-thiazol-2-yl}acetamide hydrochloride

  • MDL number MFCD18785506
  • Cas Number : 1311314-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75012

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75012
Availability details for
Quantity
  • Availability
  • Add to Cart