ENA321763076 Enamine

methyl 4-methoxypyrrolidine-2-carboxylate hydrochloride

  • MDL number MFCD18785509
  • Cas Number : 2170123-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75029
Availability details for
Quantity
  • Availability
  • Add to Cart