ENA321763079 Enamine

3-[2-(methylamino)ethyl]-2,3-dihydro-1,3-benzoxazol-2-one hydrochloride

  • MDL number MFCD18785512
  • Cas Number : 1311314-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-75046

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75046
Availability details for
Quantity
  • Availability
  • Add to Cart