ENA321763091 Enamine

2-[4-(difluoromethoxy)-3-methoxyphenyl]ethan-1-amine hydrochloride

  • MDL number MFCD18785524
  • Cas Number : 1311315-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75151

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75151
Availability details for
Quantity
  • Availability
  • Add to Cart