ENA321763097 Enamine

2-[4-(2-aminoethyl)phenyl]acetic acid hydrochloride

  • MDL number MFCD18785530
  • Cas Number : 37563-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75206
Availability details for
Quantity
  • Availability
  • Add to Cart