ENA321763102 Enamine

5-bromo-2-(chloromethyl)pyridine hydrochloride

  • MDL number MFCD09753756
  • Cas Number : 936342-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163...165
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75239

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75239
Availability details for
Quantity
  • Availability
  • Add to Cart