ENA321763106 Enamine

2-(2,3-difluorophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD18785538
  • Cas Number : 215797-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75249
Availability details for
Quantity
  • Availability
  • Add to Cart