ENA321763108 Enamine

2-[4-(benzyloxy)phenyl]cyclopropan-1-amine hydrochloride

  • MDL number MFCD18785540
  • Cas Number : 1311317-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214...216
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75259

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75259
Availability details for
Quantity
  • Availability
  • Add to Cart