ENA321763109 Enamine

1-[2-(morpholin-4-yl)ethyl]-1H-pyrazole-4-carboxylic acid hydrochloride

  • MDL number MFCD18785541
  • Cas Number : 1311313-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206...208
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75271

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75271
Availability details for
Quantity
  • Availability
  • Add to Cart