ENA321763113 Enamine

3-bromo-1-(2-fluorophenyl)pyrrolidin-2-one

  • MDL number MFCD18785545
  • Cas Number : 188533-10-0
  • Molecular Weight : 258.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75309
Availability details for
Quantity
  • Availability
  • Add to Cart