ENA321763116 Enamine

4-(4-bromothiophen-2-yl)-1,3-thiazol-2-amine

  • MDL number MFCD18838689
  • Cas Number : 81216-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75323
Availability details for
Quantity
  • Availability
  • Add to Cart