ENA321763121 Enamine

methyl 1-(3-aminopropyl)piperidine-4-carboxylate dihydrochloride

  • MDL number MFCD18785551
  • Cas Number : 1311318-00-9
  • Molecular Weight : 273.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75353
Availability details for
Quantity
  • Availability
  • Add to Cart