ENA321763124 Enamine

3-(4-aminophenoxy)propanoic acid hydrochloride

  • MDL number MFCD18785554
  • Cas Number : 1311313-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75370
Availability details for
Quantity
  • Availability
  • Add to Cart