ENA321763128 Enamine

3-aminopyrrolidine-1-carboxamide dihydrochloride

  • MDL number MFCD18838693
  • Cas Number : 1306603-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183...185
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75402

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75402
Availability details for
Quantity
  • Availability
  • Add to Cart