ENA321763131 Enamine

2-chloro-N-(4-methoxy-1,3-benzothiazol-2-yl)acetamide hydrochloride

  • MDL number MFCD18785558
  • Cas Number : 1311317-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75440

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75440
Availability details for
Quantity
  • Availability
  • Add to Cart