ENA321763136 Enamine

2-amino-3-methoxy-2-methylpropanamide hydrochloride

  • MDL number MFCD18785563
  • Cas Number : 1311316-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215...217
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75500

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75500
Availability details for
Quantity
  • Availability
  • Add to Cart