ENA321763138 Enamine

1-(2,2-dimethyl-2,3-dihydro-1-benzofuran-5-yl)propan-1-amine hydrochloride

  • MDL number MFCD18785565
  • Cas Number : 1311313-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75507
Availability details for
Quantity
  • Availability
  • Add to Cart