ENA321763139 Enamine

N1-(6-methylpyridin-2-yl)ethane-1,2-diamine dihydrochloride

  • MDL number MFCD18785566
  • Cas Number : 1311316-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75508
Availability details for
Quantity
  • Availability
  • Add to Cart