ENA321763142 Enamine

6-methoxy-7-nitro-1,2,3,4-tetrahydronaphthalen-1-one

  • MDL number MFCD18648282
  • Cas Number : 88628-54-0
  • Molecular Weight : 221.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75532
Availability details for
Quantity
  • Availability
  • Add to Cart