ENA321763144 Enamine

5-bromo-4-nitrothiophene-2-sulfonamide

  • MDL number MFCD18838696
  • Cas Number : 64729-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75542
Availability details for
Quantity
  • Availability
  • Add to Cart