ENA321763145 Enamine

2-[3-(trifluoromethyl)phenyl]cyclopropan-1-amine hydrochloride

  • MDL number MFCD18785569
  • Cas Number : 2711-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75544

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75544
Availability details for
Quantity
  • Availability
  • Add to Cart