ENA321763150 Enamine

5-(chloromethyl)-3-cyclohexyl-4,5-dihydro-1,2-oxazole

  • MDL number MFCD18838698
  • Cas Number : 1306606-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75575
Availability details for
Quantity
  • Availability
  • Add to Cart