ENA321763152 Enamine

2-amino-1-[4-(trifluoromethyl)phenyl]butan-1-ol hydrochloride

  • MDL number MFCD18838699
  • Cas Number : 1306604-18-1
  • Molecular Weight : 269.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75602

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75602
Availability details for
Quantity
  • Availability
  • Add to Cart