ENA321763155 Enamine

ethyl 2-(aminomethyl)-1,3-thiazole-4-carboxylate dihydrochloride

  • MDL number MFCD18838702
  • Cas Number : 1306603-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128...130
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75624

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75624
Availability details for
Quantity
  • Availability
  • Add to Cart