ENA321763157 Enamine

N-(1-methanesulfonylpropan-2-yl)cyclopropanamine

  • MDL number MFCD18838704
  • Cas Number : 1306603-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75639
Availability details for
Quantity
  • Availability
  • Add to Cart