ENA321763158 Enamine

N-cyclopropyl-3-(piperazin-1-yl)propanamide dihydrochloride

  • MDL number MFCD18785574
  • Cas Number : 1311318-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75648
Availability details for
Quantity
  • Availability
  • Add to Cart