ENA321763162 Enamine

2-(3-fluorophenyl)-2-(piperazin-1-yl)ethan-1-ol dihydrochloride

  • MDL number MFCD18785578
  • Cas Number : 1311318-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75660
Availability details for
Quantity
  • Availability
  • Add to Cart