ENA321763163 Enamine

2-(4-cyclobutanecarbonylpiperazin-1-yl)ethan-1-amine hydrochloride

  • MDL number MFCD18785579
  • Cas Number : 1311317-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...202
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75671

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75671
Availability details for
Quantity
  • Availability
  • Add to Cart