ENA321763164 Enamine

3-bromoimidazo[1,2-a]pyridine-2-carbaldehyde hydrobromide

  • MDL number MFCD18785580
  • Cas Number : 1311317-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75675
Availability details for
Quantity
  • Availability
  • Add to Cart