ENA321763167 Enamine

4-[5-(chloromethyl)-1,2,4-oxadiazol-3-yl]pyridine hydrochloride

  • MDL number MFCD18785583
  • Cas Number : 1311313-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184...186
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75681

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75681
Availability details for
Quantity
  • Availability
  • Add to Cart