ENA321763173 Enamine

2-(pyrrolidin-2-yl)-4-(thiophen-2-yl)-1H-imidazole dihydrochloride

  • MDL number MFCD18785589
  • Cas Number : 1311314-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75731

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75731
Availability details for
Quantity
  • Availability
  • Add to Cart