ENA321763174 Enamine

4-methoxybutanimidamide hydrochloride

  • MDL number MFCD18785590
  • Cas Number : 1311315-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-75747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75747
Availability details for
Quantity
  • Availability
  • Add to Cart