ENA321763175 Enamine

1H,4H,5H,6H,7H-[1,2,3]triazolo[4,5-b]pyridine

  • MDL number MFCD18785591
  • Cas Number : 1125405-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163...165
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75758

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75758
Availability details for
Quantity
  • Availability
  • Add to Cart