ENA321763179 Enamine

N-(2-aminoethyl)-2,6-difluorobenzamide hydrochloride

  • MDL number MFCD18785594
  • Cas Number : 1311317-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167...169
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75783

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75783
Availability details for
Quantity
  • Availability
  • Add to Cart