ENA321763180 Enamine

benzyl (3R)-3-(chlorosulfonyl)pyrrolidine-1-carboxylate

  • MDL number MFCD18785595
  • Cas Number : 1134112-00-7
  • Molecular Weight : 303.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75789

Safety Information

signalword Danger
pcodes P260,P264,P270,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P330,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-75789
pictograms GHS05,GHS07
hstatements H302,H314
Availability details for
Quantity
  • Availability
  • Add to Cart