ENA321763197 Enamine

2-(5-chloroquinolin-8-yl)acetonitrile

  • MDL number MFCD18838716
  • Cas Number : 1306606-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75847

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-75847
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart