ENA321763217 Enamine

1-[2-(methylamino)ethyl]pyrrolidin-2-one hydrochloride

  • MDL number MFCD18785607
  • Cas Number : 1311313-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75940

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75940
Availability details for
Quantity
  • Availability
  • Add to Cart