ENA321763222 Enamine

1-[(oxiran-2-yl)methyl]pyrrolidine-2,5-dione

  • MDL number MFCD00460358
  • Cas Number : 20958-21-8
  • Molecular Weight : 155.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74774

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74774
Availability details for
Quantity
  • Availability
  • Add to Cart