ENA321763224 Enamine

1-(methylamino)-3-(4-methylpiperazin-1-yl)propan-2-ol

  • MDL number MFCD00776867
  • Cas Number : 462066-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74778
Availability details for
Quantity
  • Availability
  • Add to Cart