ENA321763232 Enamine

3-(aminomethyl)cyclohexan-1-amine

  • MDL number MFCD13185923
  • Cas Number : 97087-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74792

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74792
Availability details for
Quantity
  • Availability
  • Add to Cart