ENA321763246 Enamine

methyl 3-(2,2,2-trifluoroethoxy)benzoate

  • MDL number MFCD18785474
  • Cas Number : 35453-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56...58
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74812

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74812
Availability details for
Quantity
  • Availability
  • Add to Cart