ENA321763247 Enamine

(2-phenylcyclopropyl)methanamine hydrochloride

  • MDL number MFCD18483297
  • Cas Number : 90874-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188...190
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74387

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74387
Availability details for
Quantity
  • Availability
  • Add to Cart