ENA321763250 Enamine

4-({[1,2,4]triazolo[4,3-a]pyridin-3-yl}methyl)aniline

  • MDL number MFCD09943606
  • Cas Number : 1016839-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74818

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74818
Availability details for
Quantity
  • Availability
  • Add to Cart