ENA321763267 Enamine

2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-phenylpropanoic acid

  • MDL number MFCD00439182
  • Cas Number : 62212-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168-169
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-74863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74863
Availability details for
Quantity
  • Availability
  • Add to Cart