ENA321763269 Enamine

1-(2-bromophenyl)pyrrolidin-3-amine

  • MDL number MFCD11894842
  • Cas Number : 1179944-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74865
Availability details for
Quantity
  • Availability
  • Add to Cart