ENA321763272 Enamine

4-(3-bromophenoxy)benzoic acid

  • MDL number MFCD11156620
  • Cas Number : 347173-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74396
Availability details for
Quantity
  • Availability
  • Add to Cart