ENA321763282 Enamine

N,2-dimethyl-4,5,6,7-tetrahydro-1,3-benzothiazol-7-amine

  • MDL number MFCD16677210
  • Cas Number : 70590-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74889

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74889
Availability details for
Quantity
  • Availability
  • Add to Cart