ENA321763283 Enamine

(3,4-dihydro-2H-pyran-2-yl)methanamine

  • MDL number MFCD00023814
  • Cas Number : 4781-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74307
Availability details for
Quantity
  • Availability
  • Add to Cart